Would you like to log in again?
Mouse Anti-body Capture Kit, Type-2, Cytiva
Supplier: Cytiva
29215281
103568-402EA 1205.77 USD InStock
103568-402
Mouse Anti-body Capture Kit, Type-2, Cytiva
Assays Immunological Assays
Kit with reagents for the capture of mouse antibodies, in component volumes larger than the standard mouse antibody capture kit.
  • Binds all IgG subclasses (IgG1, IgG2a, IgG2b, and IgG3), IgA and IgM
  • Very stable binding with no or very low dissociation of captured antibodies
Designed for use with Biacore, but can also be used with all other Biacore systems.
Mouse antibody capture kit, type 2 is intended for capture of mouse antibodies as ligands in various biomolecular interaction analyses. Anti-mouse antibodies are suitable for immobilization on carboxyl derivatized surfaces using amine coupling kit and the included immobilization buffer, while the regeneration solution is used for regeneration of the surface.
Anti-mouse antibodies consist of polyclonal rabbit anti-mouse immunoglobulin antibodies reacting with all IgG subclasses.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty