Would you like to log in again?
His Capture Kit, Type 2, Cytiva
Supplier: Cytiva
29234602
103568-410EA 1923.38 USD InStock
103568-410
His Capture Kit, Type 2, Cytiva
Assays Protein Assays
Reagents for capture of histidine-tagged molecules as ligands in biomolecular interaction analysis. Designed for use with Biacore 8K+, Biacore 8K (larger kit component volumes than in His Capture Kit) but can be used with all other Biacore systems.
  • Enables capture of histidine-tagged ligands
  • Physiological conditions can be used during the coupling
  • Optimized and verified assay minimizing assay development
It is designed for capture of histidine-tagged proteins to an anti-histidine antibody and provides an alternative to capture on nickel-chelated nitrilotriacetic (NTA) groups.
The anti-histidine antibody is a monoclonal antibody directed against polyhistidine tags in the C- or N-terminus of a protein. Depending on the nature of the interacting molecules and the micro-environment around the histidine-tag, the affinity of the capture antibodies will vary.
Delivery information: Kit contains: Anti-histidine antibody 1 mg/ml in 0.15 M NaCl, 90 μl; Immobilization buffer 10 mM sodium acetate pH 4.5, 2.6 ml; Regeneration solution: 10 mM glycine-HCl, pH 1.5, 2×120 ml.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty