Would you like to log in again?
Horseradish Peroxidase (HRP) Chromogen Substrates for IHC, Vector Laboratories
Supplier: Vector Laboratories
SK-4103-400 SK-4400 SK-4600 SK-4285 SK-4800 SK-4605 SK-4700 SK-4200 SK-4805 SK-4105 SK-4705 SK-4205 SK-4100
103572-370EA 264.46 USD InStock
103572-370 103572-372 101098-446 101098-436 101098-444 101098-434 101098-440 103383-922 103572-366 103572-378 103572-376 101098-448 103572-368
Horseradish Peroxidase (HRP) Chromogen Substrates for IHC, Vector Laboratories
Blotting Kits
Vector Laboratories horseradish peroxidase (HRP) substrate kits include options that are cost-effective (e.g. standard DAB), sensitive (e.g ImmPACT DAB) and convenient (ImmPACT DAB EqV). The range of vibrant colors facilitates multiplex staining and improved contrast with pigmented tissues and counterstains.
  • Optimized for IHC, ICC, ISH and blots
  • Stock solutions are supplied in convenient dropper bottles for greater ease of handling
  • Compatible with either VectaMount Permanent Mounting Media (H-6000) or VectaMount AQ Aqueous Mounting Media (H-5501)
  • One year expiry date
For immuno-visualization experiments to be successful, one needs proper sample prep, a well-targeted primary anybody, an effective secondary antibody-based detection system and a high-quality chromogenic substrate. Vector Laboratories has decades of experience offering exceptional staining systems and substrates.
Standard substrates provide exceptional value and convenience over substrates in the form of powders or pills and work well for higher-abundance targets. Vector Laboratories ImmPACT substrates offer up to five times higher sensitivity with convenient handling and are ideal for detecting lower-abundance targets.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty