Would you like to log in again?
Lactoferrin ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
KR6870
103573-008EA 1170.38 USD InStock
103573-008
Lactoferrin ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
The Lactoferrin ELISA is intended for the determination of lactoferrin in stool.
  • Sensitivity: 0.369 ng/ml
  • Dynamic Range: 2.4 to 240 ng/ml
  • Sample Size: 100 μl
  • Sample Type: Serum
  • Incubation Time: 3.5 hours
Lactoferrin is a 76 kDa iron-binding glycoprotein which is synthesised and stored in the secondary granules of neutrophils. It is also present in several secretory fluids, such as milk, saliva, tears, and nasal secretions.
Lactoferrin can exist in different polymeric forms ranging from monomers to tetramers; it tends to polymerise especially at high concentrations. The physiological activities of lactoferrin include regulation of iron homeostasis, innate defense against a broad range of microbial infections, anti-inflammatory activity, regulation of cellular growth and differentiation and protection against cancer development and metastasis.
During intestinal inflammation, polymorphonuclear neutrophils infiltrate the mucosa and release lactoferrin by degranulation, which results in an increased excretion of lactoferrin into the faeces. Faecal lactoferrin is therefore a marker for neutrophilic intestinal inflammation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty