Would you like to log in again?
MICRO TUBE STANDARD PP YL 1.5ML PK500
Supplier: SARSTEDT INC MS
Standard micro tube 1.5ml^ Standard micro tube, Material: Polypropylene, Color: Yellow, With flat cap, Wide hinge for easy and safe closure, Large writing space, Centrifugation: 20,000xg, Size: 1.5ml
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MICRO TUBE STANDARD PP YL 1.5ML PK500 103575-320 72.690.378 Pack of 500 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty