Would you like to log in again?
AGAROSE BEAD RFP-TRAP MAGNETIC 100 RXNS
Supplier: Bulldog Bio
RFP Trap Coupled to Magnetic Agarose Bead 100 reactions^ RFP Trap Coupled to Magnetic Agarose Bead, 2.5 ml; Binding Capacity: 10ul binds 3-4ug of recombinant RFP, Particle Size: approx 40 um, Storage Buffer: 20% EtOH, for immuno-purification and immuno-pull down experiments, Size: 100 reactions
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
AGAROSE BEAD RFP-TRAP MAGNETIC 100 RXNS 103575-326 RTMA100 Each (2,500µl) Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty