Would you like to log in again?
AGAROSE BEAD KIT RFP-TRAP MAGNETIC 20RXN
Supplier: Bulldog Bio
RFP-Trap magnetic agarose bead kit with lysis 20 rxns^ RFP-Trap, coupled to magnetic agarose bead, kit with lysis, wash and elution buffers, Binding Capacity: 10ul binds 3-4ug of recombinant RFP, Particle Size: approx 40 um, Storage Buffer: 20% EtOH,for immuno-purification, Size: 20 reactions
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
AGAROSE BEAD KIT RFP-TRAP MAGNETIC 20RXN 103575-344 RTMAK020 Kit of 1 Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty