Would you like to log in again?
SpongeCol® Type I Collagen Sponge, Advanced BioMatrix
Supplier: Advanced BioMatrix
5135-5EA 5135-25EA
103700-662PK 549.23 USD InStock
103700-662 103700-660
SpongeCol® Type I Collagen Sponge, Advanced BioMatrix
Cell Culture Supplements and Additives
Advanced BioMatrix products are recognized for purity, functionality, quality, and lot-to-lot consistency, allowing for reproducibility in research.
  • Columnar pore architecture type I collagen sponge
  • Lightly crosslinked for durability
  • Great for tissue engineering
SpongeCol® is a collagen sponge with an interpenetrating, columnar porous architecture structure. SpongeCol® contains unique columnar porous network which permits cells and nutrients to flow completely through the interpenetrating pores and provides an increased surface area for cell attachment, growth and migration.
SpongeCol® is composed of highly purified type I collagen which best supports the attachment, proliferation, and function of cells. The collagen is lightly cross-linked for increased mechanical strength and durability for short- and long-term tissue culture yet is still biodegradable over the longer-term. The diameter of the pores ranges from 100 to 400 micron with the average diameter being approximately 200 microns. Each collagen disc is approximately 4 mm in diameter and 1.5 mm thick. The sponge discs fits into a 96- well culture plate well. This product is terminally sterilized and ready-to-use.
Caution: This product is for R and D use only and is not intended for human or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty