Would you like to log in again?
Collagenase
Supplier: Advanced BioMatrix
5030-50MG
103701-190EA 104.62 USD InStock
103701-190
Collagenase
Enzymes
Collagenase consists of a blend of enzymes including collagenase, caseinase, clostripain and trypsin designed to effectively hydrolyze collagen and dissociate tissues. Various collagenase products have been evaluated specifically for use with Advanced BioMatrix’ s collagen products. This product is offered to improve the viability and functionality of isolated cells from the digested collagen gels.
Collagenase is provided as a lyophilized, sterile powder in a 50 mg package size. After reconstitution, the product is ready-to-use.
  • Digest 3D collagen hydrogels
  • Lyophilized and ready to use
  • Sterile powder
Caution: This product is for R&D use only and is not intended for human or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty