Would you like to log in again?
Phosphate Buffered Saline, 10X, Advanced Biomatrix
Supplier: Advanced BioMatrix
5076-100ML
103701-212EA 85 USD InStock
103701-212
Phosphate Buffered Saline, 10X, Advanced Biomatrix
Buffers Biological Buffers
Use With Type I Collagen to help form 3D hydrogels.
  • Sterility tested
  • Useful for 3D collagen hydrogel procedure
  • Sterile filtered
  • pH: Approx pH 7.0 to 7.6
Formulated to use in conjunction with other products such as Type I collagen for forming a gel.
PBS 10X is provided in a user-friendly packaging for use and storage. This product is sterile and supplied as a ready to use solution.
Caution: This product is for R&D use only and is not intended for human or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty