Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 0.01M, Sterile-Filtered

Supplier: ADVANCED BIOMATRIX, INC. MS
Danger

5077-100ML
103701-214EA 85 USD InStock
103701-214
Hydrochloric acid 0.01M, Sterile-Filtered
Hydrochloric acid
HCl is provided in a user-friendly packaging for use and storage. This product is sterile and supplied as a ready to use solution.

  • Sterility tested
  • Ready-to-use solution
  • Approx pH 1.5 to 2.5
  • For Use in Preparation of 3D Matrix Products

Hydrochloric Acid (HCl) 0.01M is formulated and prepared to use in conjunction with other products provided by Advanced BioMatrix.


Formula: HCl
MW: 36.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty