Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 0.1M, sterile-filtered

Supplier: ADVANCED BIOMATRIX, INC. MS
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

5078-100ML
103701-216EA 85 USD InStock
103701-216
Sodium hydroxide 0.1M, sterile-filtered
Sodium hydroxide
NaOH is provided in a user-friendly packaging for use and storage. This product is sterile and supplied as a ready to use solution.

  • Sterility tested
  • Ready-to-use solution
  • Approx pH 11.5 to 13.5
  • Useful for 3D collagen hydrogel procedure

Sodium Hydroxide (NaOH) Solution 0.1M is formulated and prepared to use in conjunction with other products provided by Advanced BioMatrix.


Formula: NaOH
MW: 40 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty