Would you like to log in again?
Purple-Jelley Hyaluronan Assay Large Economy Kit, BioVendor
Supplier: BioVendor
H2000
103737-126EA 2457.75 USD InStock
103737-126
Purple-Jelley Hyaluronan Assay Large Economy Kit, BioVendor
Assays Cellular Assays
The Purple-Jelley hyaluronan assay is a quantitative assay used to measure the quantity of hyaluronic acid in mammalian tissue.
  • Regulatory status: RUO
  • Type: colorimetric assay
  • Species: multispecies
Hyaluronic acid (HA), also known as hyaluronan, is a non-sulphated glycosaminoglycan (GAG) and is composed of repeating polymeric disaccharides of D-glucuronic acid and N-acetyl-D-glucosamine linked by a glucuronidic β (1→3) bond. In aqueous solutions HA forms specific stable tertiary structures. Despite the simplicity in its composition, without variations in its sugar composition or without branching points, HA has a variety of physicochemical properties. HA polymers occur in a vast number of configurations and shapes, depending on their size, salt concentration, pH, and associated cations. Unlike other GAG, HA is not covalently attached to a protein core, but it may form aggregates with proteoglycans. HA encompasses a large volume of water giving solutions high viscosity, even at low concentrations.
HA is widely distributed, from prokaryotic, to eukaryotic cells. In humans, HA is most abundant in the skin, (accounting for 50% of the total body HA), the vitreous of the eye, the umbilical cord, and synovial fluid, but it is also present in all tissues and fluids of the body, such as skeletal tissues, heart valves, the lung, the aorta, the prostate, tunica albuginea, corpora cavernosa and corpus spongiosum of the penis. HA is produced primarily by mesenchymal cells but also by other cell types.
Considerable evidence exists regarding the functional role of HA in molecular mechanisms and, consequently, the potential role of HA for the development of novel therapeutic strategies for many diseases.
Functions of HA include the following: hydration, lubrication of joints, a space filling capacity, and the framework through which cells migrate. The synthesis of HA increases during tissue injury and wound healing and HA regulates several aspects of tissue repair, including activation of inflammatory cells to enhance immune response and the response to injury of fibroblasts and epithelial cells. HA also provides the framework for blood vessel formation and fibroblast migration, that may be involved in tumor progression. The correlation of HA levels on the cell surface of cancer cells with the aggressiveness of tumors has also been reported.
The size of HA appears to be of critical importance for its various functions described above. HA of high molecular size, usually in excess of 1000 kDa, is present in intact tissues and is antiangiogenic and immunosuppressive, whereas smaller polymers of HA are distress signals and potent inducers of inflammation and angiogenesis.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty