Would you like to log in again?
Chorionic Gonadotropin Human
Supplier: BioVendor
RP17825010 RP17825005
103737-130EA 895.92 USD InStock
103737-130 103737-128
Chorionic Gonadotropin Human
Proteins and Peptides
Human Chorionic Gonadotropin is produced from a sterile preparation of placental glucoprotein urine of pregnant women having a total molecular mass of 36700 Dalton.
  • Urine of pregnant women
  • Sterile Filtered White lyophilized (freeze-dried) powder
  • The hCG was lyophilized with no additives
  • It is recommended to reconstitute the lyophilized Human Chorionic Gonadotropin in sterile 18 MΩ-cm
  • H₂O not less than 100 µg/ml, which can then be further diluted to other aqueous solutions
The hCG consists of 237 amino acids, a chain-92 amino acids and b chain-145 amino acids. The hCG is purified by proprietary chromatographic techniques.
Caution: Research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty