Would you like to log in again?
Apoptosis Assay Cell Apopercentage, BioVendor
Supplier: BioVendor
A1000
103737-158EA 740.09 USD InStock
103737-158
Apoptosis Assay Cell Apopercentage, BioVendor
Assays Cellular Assays
The Cell-APOPercentage™ apoptosis assay is a detection and measurement system to monitor the occurrence of apoptosis in mammalian, anchorage-dependent cells during in vitro culture.
  • Regulatory status:RUO
  • Type: cell-based assay
  • Species: multispecies
The onset of the execution phase of apoptosis has been linked to translocation of phosphatidylserine from the interior to the exterior surface of the mammalian cell membrane, experimentally supported by annexin-v binding to phosphatidylserine. Phosphatidylserine transmembrane movement results in the uptake of the APOPercentage dye by apoptotic committed cells. This dye uptake continues until blebbing occurs. No dye is then released from the now defunct cell, other than that within isolated ‘blebs’. Each apoptotic inducer has an optimum time and concentration to be effective. This optimum time and concentration will be affected by the cell line examined.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty