Would you like to log in again?
Sircol™ Soluble and Insoluble Collagen, BioVendor
Supplier: BioVendor
S4000 S6000 S2000
103737-186EA 1589.25 USD InStock
103737-186 103737-182 103737-184
Sircol™ Soluble and Insoluble Collagen, BioVendor
Assays Cellular Assays
Collagen is the most abundant protein found in animals. During a healthy life span the insoluble, covalent, cross-linked collagen fibers retain their biophysical functions and shapes. The collagen fibers remain isolated from most of the biochemical activities of the cells.
  • Dye-binding method
  • It replaces the conventional hydroxyproline analysis. No more working with concentrated acids or alkalis
  • The Sircol Insoluble Assay uses mild acid and temperature treatment for 2 to 3 hrs
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty