Would you like to log in again?
Urine Pre-Clinical CartiLaps® ELISA (CTX-II), BioVendor
Supplier: BioVendor
AC-09F1
103739-554EA 2124.31 USD InStock
103739-554
Urine Pre-Clinical CartiLaps® ELISA (CTX-II), BioVendor
Assays ELISAs
96 wells (1 kit) - ELISA is a competitive enzyme immunoassay for the quantitative measurement of CTX-II.
The Urine Pre-Clinical CartiLaps® (CTX-II) EIA detects degradation products of C-terminal telopeptides of type II collagen (CTX-II) in non-human urine and cell culture supernatant. Background - Disruption of the structural integrity of cartilage is the major histological finding in osteoarthritis and rheumatoid arthritis. Type II collagen is the major organic constituent of cartilage and fragments of type II collagen (CTX-II) are being released into circulation and subsequently secreted into urine following degradation of cartilage. In non-human urine, the CTX-II fragments can be quantified by Urine Pre-Clinical CartiLaps® (CTX-II) EIA, which in addition is applicable for cell culture supernatants. The corresponding test for human application, i.e. the Urine Pre-Clinical CartiLaps® (CTX-II) EIA, has been reported to be useful in prediction of progression of osteoarthritis (Reijman (2003), Garnero (2003)) and in other clinical investigations (please refer to REFERENCES).
The Urine Pre-Clinical CartiLaps® (CTX-II) EIA is a modification of the Urine CartiLaps® EIA (Christgau (2001), which is based on the competitive binding of a monoclonal antibody to urinary fragments of type II collagen or to biotinylated, synthetic peptides bound to the surface of microtitre plates coated with streptavidin. The modification allows a broadening of the measuring range facilitating the measurement of non-human urines having a wide range of CTX-II values. Initially, biotinylated, synthetic peptides are bound to the surface of streptavidin-coated wells of the microtitre plate. After washing, standards, controls and urine samples are pipetted into the wells followed by addition of a solution of the monoclonal antibody. The wells are washed, and a solution of peroxidase-conjugated anti-mouse immunoglobulin (rabbit) is added to the wells. Following the second washing step, a chromogenic substrate is added to all wells and the colour reaction is stopped with sulphuric acid and the absorbance is measured.
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty