Would you like to log in again?
Urine CartiLaps® ELISA (CTX-II), BioVendor
Supplier: BioVendor
AC-10F1
103739-556EA 2012.66 USD InStock
103739-556
Urine CartiLaps® ELISA (CTX-II), BioVendor
Assays ELISAs
96 wells (1 kit) - ELISA is a competitive enzyme immunoassay for the quantitative measurement of CTX-II.
The Urine CartiLaps® (CTX-ll) EIA detects degradation products of C-terminal telopeptides of type II collagen. Background - Disruption of the structural integrity of cartilage is the major histological finding in osteoarthritis and rheumatoid arthritis. Type II collagen is the major organic constituent of cartilage and fragments of type II collagen (CTX-II) are being released into circulation and subsequently secreted into urine following degradation of cartilage. In urine, the CTX-II fragments can be quantified by Urine CartiLaps® (CTX-ll) EIA. The Urine CartiLaps® (CTX-ll) EIA has been reported to be useful in prediction of progression of osteoarthritis (Reijman (2003), Garnero (2003)) and in other clinical and pre-clinical investigations (please refer to REFERENCES).
Urine CartiLaps® (CTX-ll) EIA (Christgau (2001) is based on the competitive binding of a monoclonal antibody to urinary fragments of type II collagen or to biotinylated, synthetic peptides bound to the surface of microtitre plates coated with streptavidin. Initially, biotinylated, synthetic peptides are bound to the surface of streptavidin-coated wells of the microtitre plate. After washing, standards, controls, and urine samples are pipetted into the wells followed by addition of a solution of the monoclonal antibody. The wells are washed, and a solution of peroxidaseconjugated anti-mouse immunoglobulin (rabbit) is added to the wells. Following the second washing step, a chromogenic substrate is added to all wells and the color reaction is stopped with sulphuric acid and the absorbance is measured.
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty