Would you like to log in again?
N-MID® Osteocalcin ELISA, Biovendor
Supplier: BioVendor
AC-11F1
103739-558EA 1140.43 USD InStock
103739-558
N-MID® Osteocalcin ELISA, Biovendor
Assays ELISAs
96 wells (1 kit) - ELISA is a sandwich enzyme immunoassay for the quantitative measurement of N-MID Osteocalcin.
The N-MID® Osteocalcin ELISA is based upon the application of two highly specific monoclonal antibodies (Mabs) against human osteocalcin. An antibody recognizing the mid region (amino acids 20 to 29) is used as the capture antibody and for detection a peroxidase conjugated antibody recognizing the N-terminal region (amino acids 10 to 16) is used. In addition to intact osteocalcin (amino acid 1 to 49) the N-terminal-Mid fragment (amino acids 1 to 43) is also detected. Standards, control and unknown samples are pipetted into the appropriate microtiter wells coated with streptavidin. Then a mixture of a biotinylated antibody and a peroxidase conjugated antibody is added. Following incubation for 2 hours at room temperature the wells are washed and a chromogenic substrate is added and the color reaction is stopped with sulfuric acid. Finally, the absorbance is measured.
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty