Would you like to log in again?
Rat-MID™ Osteocalcin ELISA, BioVendor
Supplier: BioVendor
AC-12F1
103739-560EA 1280 USD InStock
103739-560
Rat-MID™ Osteocalcin ELISA, BioVendor
Assays ELISAs
96 wells (1 kit) - ELISA is a sandwich enzyme immunoassay for the quantitative measurement of osteocalcin.
The Rat-MID™ Osteocalcin EIA is an enzyme-linked immunosorbent assay for the quantitative determination of osteocalcin in rat serum and plasma. The assay is for research-use-only. Summary and explanation of the test - Osteocalcin or bone Gla protein (BGP), is a major non-collagenous protein of the bone matrix. It has a molecular weight of approximately 6000 Dalton and consists in most species of 49 amino acids; however, rat osteocalcin consist of 50 amino acids (1-4). Osteocalcin has a unique property for binding to calcium facilitated by the presence of 2-3 gamma-carboxyglutamic acids at position 17, 21 and 24 (5). The mid-molecular part of osteocalcin, especially the amino acids between 20 and 30, exhibits a high degree of inter-species conservatism (4).
The Rat-MID™ Osteocalcin EIA is based upon the competitive binding of a monoclonal antibody to soluble osteocalcin or to immobilized osteocalcin. Briefly, the monoclonal antibody is raised against human osteocalcin and recognizes the mid-molecular part (amino acids 21-29) of the molecule (11). Synthetic human osteocalcin is used for standardization. Parallelism is observed with purified rat osteocalcin and rat serum. During the Pre-incubation step, biotinylated osteocalcin is immobilized by binding to the streptavidin-coated microtitre wells. The wells are emptied and washed, and standards, control, or unknown serum samples are pipetted into appropriate wells, followed by a solution of monoclonal antibody. Following the primary-incubation step the wells are emptied and washed. In the Secondary-incubation step a solution of peroxidase conjugated anti-mouse immunoglobulins is added and binds to the monoclonal antibody. After the third washing step a chromogenic substrate (TMB) is added and the color reaction is stopped with sulfuric acid. Finally, the absorbence at 450 nm is measured. The absorbence level is inversely related to concentration of osteocalcin in the sample
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty