Would you like to log in again?
1,25(OH)₂ Vitamin D Total ELISA, BioVendor
Supplier: BioVendor
RIS021R
103739-616EA 863.92 USD InStock
103739-616
1,25(OH)₂ Vitamin D Total ELISA, BioVendor
Assays ELISAs
96 wells (1 kit) - ELISA is a competitive enzyme immunoassay for the quantitative measurement of 1,25(OH)₂ Vitamin D.
Immunoenzymetric assay for the quantitative measurement of 1,25(OH)₂ Vitamin D in serum.
Vitamin D is mainly synthesized in the skin from 7-dehydrocholesterol and is partially from dietary and supplementation origin. In the liver, Vitamin D is hydroxylated on carbon 25 to produce the intermediate 25OH Vitamin D. 25OH Vitamin D is further metabolized before it can carry out the functions of Vitamin D on intestine, kidneys, bone and other organs and tissues.
Only samples and controls, not the calibrators, are extracted with a mixture of solvents and applied on cartridges to separate 1,25(OH)₂ Vitamin D from the other Vitamin D metabolites. After elution of the 1,25(OH)₂ Vitamin D from the samples and controls cartridges, the calibrators, eluted samples and eluted controls are incubated directly in microtiterplate coated with anti1,25(OH)₂ Vitamin D antibodies. After an overnight incubation at 4 °C, the microtiter plate is washed and the working conjugate solution is added and incubated for 1 hour at 4 °C. The microtiterplate is then washed to stop the competition reaction. The Chromogenic solution (TMB) is added and incubated for 15 minutes at room temperature (18 to 25 °C). The reaction is stopped with the addition of Stop Solution and the microtiterplate is read at the appropriate wavelength. The amount of 1,25(OH)₂ Vitamin D is determined colorimetrically by measuring the absorbance, which is inversely proportional to the 1,25(OH)₂ Vitamin D concentration. A calibration curve is plotted and the 1,25(OH)₂ Vitamin D concentrations of the samples are determined by dose interpolation from the calibration curve.
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty