Would you like to log in again?
ENDONEXT™ EndoZyme® II Go Fast Recombinant Factor C (rFC) Endotoxin Detection Assay, BioVendor
Supplier: BioVendor
890031
103739-724EA 2025.23 USD InStock
103739-724
ENDONEXT™ EndoZyme® II Go Fast Recombinant Factor C (rFC) Endotoxin Detection Assay, BioVendor
Assays Enzyme Assays
The ENDONEXT™ EndoZyme® II assay is the enhanced second generation of EndoZyme®, based on recombinant Factor C (rFC), the synthetic analogue to the native endotoxin-specific principal receptor in the limulus amebocyte lysate (LAL) cascade.
  • Assay run time: 20 minutes (0.05 EU/ml), 60 minutes (0.005 EU/ml)
  • Preloaded microplate eliminates manual preparation of standard dilutions and PPCs
  • Over 50% reduction in handling time compared to conventional microplate assays
  • Significantly reduces risk of handling error during preparation steps
  • Higher precision and rate of valid results ideal for in-process control of water and raw materials, as well as product release testing ideal for automation and parallel processing of plates
Instead of being based on an animal source, EndoZyme® II is a sustainable alternative method in line with regulatory requirements.
The rapid GO version of EndoZyme® II features GOPLATE™ - a microplate pre-filled with dried control standard endotoxin, standard curve 0.005 - 50 EU/ml and positive product Ccontrols (PPC) 0.5 EU/ml, all in duplicate replicates fulfilling pharmacopoeia standard curve requirements.
This new version of EndoZyme® enables endotoxin testing in pharmaceutical grade water, injectable drugs and other pharmaceutical products. EndoZyme® II GO allows an easy and fast workflow, and is particularly adapted to raw materials and final product testing.
Ships at ambient temperature. Upon receipt, store the product at the temperature recommended below. Unopened kits are stable at 2...8°C until the expiry date printed on the label. The calibration range is 0.005 – 50 EU/ml. The limit of detection is 0.005 EU/ml.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty