Would you like to log in again?
CrossLaps® for Culture ELISA (CTX-I), BioVendor
Supplier: BioVendor
AC-07F1
103739-730EA 1719.58 USD InStock
103739-730
CrossLaps® for Culture ELISA (CTX-I), BioVendor
Assays ELISAs
96 wells (1 kit) - ELISA is a competitive enzyme immunoassay for the quantitative measurement of CTX-I.
The test is an enzyme-linked immunosorbent assay (ELISA) for the quantitative determination of degradation products of C-terminal telopeptides of type I collagen in bone cell culture supernatants. It represents a technically improvement of the assay originally described in 1996 (6). Summary and explanation of the test - Type I collagen accounts for more than 90% of the organic matrix of bone (1). During renewal of the skeleton bone matrix is degraded and consequently fragments of type I collagen is released into circulation. The resorption process can be studied in vitro by culturing bone cells on devitalised slices of bone or dentin.
The assay is based on the simultaneous binding of the collagen fragments by two monoclonal antibodies, one of which will subsequently mediate the binding to the solid surface of the microtitre plate. Standards, control, or unknown samples are pipetted into the appropriate well of a microtitre plate coated with streptavidin, and subsequently a mixture of a biotinylated antibody and a peroxidase-conjugated antibody is added. Then a complex between the collagen fragments and the two antibodies is generated, and the complex will bind to the streptavidin surface via the biotinylated antibody. Following this one step incubation at room temperature the wells are washed and a chromogenic substrate added. The color reation is stopped with sulphuric acid and the absorbance is measured.
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty