Would you like to log in again?
Interleukin-18 Mouse ELISA, BioVendor
Supplier: BioVendor
RAF144R
103739-750EA 1014.71 USD InStock
103739-750
Interleukin-18 Mouse ELISA, BioVendor
Assays ELISAs
96 wells (1 kit) - ELISA is a competitive enzyme immunoassay for the quantitative measurement of IL-18.
The mouse IL-18 ELISA is an enzyme-linked immunosorbent assay for the quantitative detection of mouse IL-18. Interleukin (IL) -18 is a newly discovered cytokine, structurally similar to IL-1, with profound effects on T-cell activation. Formerly called interferon (IFN) gamma inducing factor (IGIF), IL-18 is a novel cytokine that plays an important role in the T-cell-helper type 1 (Th1) response, primarily by its ability to induce IFN gamma production in T cells and natural killer (NK) cells. An anti-mouse IL-18 coating antibody is adsorbed onto microwells.
Mouse IL-18 present in the sample or standard binds to antibodies adsorbed to the microwells. A biotinconjugated anti-mouse IL-18 antibody is added and binds to mouse IL-18 captured by the first antibody. Following incubation unbound biotin-conjugated anti-mouse IL-18 antibody is removed during a wash step. Streptavidin-HRP is added and binds to the biotin-conjugated anti-mouse IL-18 antibody. Following incubation unbound Streptavidin-HRP is removed during a wash step, and substrate solution reactive with HRP is added to the wells. A colored product is formed in proportion to the amount of mouse IL-18 present in the sample or standard. The reaction is terminated by addition of acid and absorbance is measured at 450 nm. A standard curve is prepared from 7 mouse IL-18 standard dilutions and mouse IL-18 sample concentration determined.
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty