Would you like to log in again?
Plastic Vials, Cytiva
Supplier: Cytiva
29266981
103743-742EA 170.23 USD InStock
103743-742
Plastic Vials, Cytiva
Vials
These PP vials are suitable for use with Biacore SPR systems.
For use with Biacore T200, Biacore S200, Biacore X100, Biacore 3000, and Biacore C SPR systems.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty