Would you like to log in again?
DH10B Chemically Competent Cells, Intact Genomics
Supplier: Intact Genomics
1018-96 1011-24 1014-24 1011-12 1014-48
103753-072EA 190.38 USD InStock
103753-072 103753-070 103753-076 103753-074 103753-068
DH10B Chemically Competent Cells, Intact Genomics
Competent Cells
10B chemically competent cells (E. coli) are suitable for high efficiency transformation in a wide variety of applications such as cloning and sub-cloning. This derivative of DH10B provides the highest efficiency in the industry.
  • DH10B chemically competent cells provide the highest transformation efficiency (≥1×1010 Cfu/µg pUC19 DNA) in the industry
  • Available in a wide variety of sizes, including single-use vials, 200 μl vials and 96 well plate
  • Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
  • Includes ig™ 10B competent cells, pUC19 control DNA and recovery medium
Genomic features a Φ80lacZΔM15 marker provides α-complementation of the β-galactosidase gene with blue/white screening. mcrA genotypic marker and the mcrBC, mrr deletion allows for cloning DNA that contains methylcytosine and methyladenine.
Delivery information: All our products are shipped with dry ice.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty