Would you like to log in again?
BL21 (DE3) Chemically Competent Cells, Intact Genomics
Supplier: Intact Genomics
1054-24 1051-12 1054-48 1051-24
103753-090EA 155.77 USD InStock
103753-090 103753-094 103753-092 103753-096
BL21 (DE3) Chemically Competent Cells, Intact Genomics
Competent Cells
Chemically competent BL21(DE3) E. coli cells are suitable for transformation and routine protein expression.
  • BL21 (DE3) chemically competent cells provide the highest transformation efficiency (≥1×109 Cfu/µg pUC19 DNA) in the industry
  • Available as single-use vials or 200 μl vials
  • Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
  • Includes BL21 (DE3) competent cells, pUC19 control DNA and recovery medium
Genomic features a widely used host background, T7 expression strain, deficiency in both lon and ompT proteases, resistance to phage T1 (fhuA2) and protease deficient B strain.
Delivery information: All our products are shipped with dry ice.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty