Would you like to log in again?
igMAX™ DH10B Electrocompetent Cells, Intact Genomics
Supplier: Intact Genomics
1284-24
103753-112EA 421.15 USD InStock
103753-112
igMAX™ DH10B Electrocompetent Cells, Intact Genomics
Competent Cells
igMax™ DH10B derivative electro-competent cells are suitable for demanding cloning situations, such as synthetic bio-applications, BAC cloning, assembling large and multi-DNA fragments, and large construct transformation or cloning difficult targets, requiring the greatest number of transformants possible. Utilizing proprietary manufacturing methods, these cells allow for effective transformation of all large DNA molecules (≥10 kb to 350 kb).
  • Extremely large DNA molecules can effectively be transformed at a rate much higher than ANY competitor
  • DH10B MAX electro-competent provide the highest transformation efficiency (≥5×1010 cfu/µg pUC19 DNA) in the industry
  • Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
  • igMax™ DH10B electro-competent cells, pUC19 control DNA and recovery medium
Genomic features a ≥5×1010 cfu/µg efficiency with electroporation, a 5 to 10×107 for 100 to 150 kb large DNA, high-efficiency transformation with plasmids 30 kb in size blue/white screening of recombinant clones due to lacZΔM15.
Delivery information: All our products are shipped with dry ice.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty