Would you like to log in again?
SS320 and TG1 Phage Display Electrocompetent Cells, Intact Genomics
Supplier: Intact Genomics
1264-24 1274-48 1264-48 1274-24
103753-128EA 478.85 USD InStock
103753-128 103753-126 103906-458 103753-132
SS320 and TG1 Phage Display Electrocompetent Cells, Intact Genomics
Competent Cells
Intact Genomics TG1 Phage Display Electrocompetent Cells are suitable for protein expression and preparation of antibody or peptide phage display libraries.
  • Intact Genomics (IG) Phage Display Competent Cells Provide the highest transformation efficiency (≥4×10¹⁰ Cfu/µg pUC19 DNA) in the industry
  • Amber suppressor strain (supE)
  • Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
Intact Genomics SS320 (MC1061F\’) Phage Display Electrocompetent Cells are suitable for protein expression, general cloning, blue/white screening, M13 phage work and phage display protein expression.
Product Includes SS320 Phage Display electrocompetent cells, pUC19 control DNA and a recovery medium.
Delivery information: All our products are shipped with dry ice via FedEx overnight delivery. US orders received by 1:00 pm (central time, monday-thursday) will typically be shipped on the same day in the US. Orders after 1:00 PM will be shipped on the next business day. Orders received on friday will be ship on the following monday. The shipping and dry ice packaging cost is $45 for your entire order in the 48 contiguous United States.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty