Would you like to log in again?
Taq DNA Polymerase
Supplier: Intact Genomics
3245 3245D
103753-146EA 686.54 USD InStock
103753-146 103753-144
Taq DNA Polymerase
Enzymes
High quality Taq DNA polymerase useful for routine PCR applications.
  • Useful for routine PCR, primer extension, colony PCR and more
  • Formulated for amplifying long target DNA
  • Efficient for amplifying high GC template DNA with magic enhancer
  • Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product.
  • Taq DNA polymerase
  • 10X PCR buffer with Mg²⁺
  • 5X Magic enhancer
Taq DNA polymerase is a thermostable DNA polymerase that possesses a 5' - 3' polymerase activity and a 5' flap endonuclease activity.
Delivery information: This product is supplied with 10X PCR reaction buffer, containing MgCl₂, which produces a final Mg²⁺ concentration of 1.5 mM. Ideal for primary extension reaction with DNA fragments having dA overhang on 3' ends. All our products are shipped with dry ice. 103753-146 includes dNTP.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty