Would you like to log in again?
igScript™ Reverse Transcriptase (from Moloney murine leukemia virus)
Supplier: Intact Genomics
3344
103753-170EA 288.46 USD InStock
103753-170
igScript™ Reverse Transcriptase (from Moloney murine leukemia virus)
Enzymes
igScript™ reverse transcriptase, is a recombinant, Moloney Murine Leukemia virus (MMLV) reverse transcriptase with reduced RNase H activity.
  • Recombinant MMLV reverse transcriptase with greatly reduced RNase H activity
  • Active at temperatures up to 55 °C
  • Highly efficient at producing full-length cDNA from as little as 50 pg of total RNA
  • Useful for first strand cDNA synthesis for PCR or RT-PCR and gene expression data validation by using RT-PCR
  • Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
  • 10X igScript™ RT reaction buffer
igScript™ reverse transcriptase (igScript RT) is a recombinant Moloney Murine Leukemia Virus (MMLV) reverse transcriptase with reduced RNase H activity. It is highly efficient at producing full-length cDNA from long RNA templates. igScript RT is an RNA-directed DNA polymerase which lacks 3' - 5' exonuclease activity and is capable of producing cDNA from as little as 50 pg of total RNA for real-time RT-PCR analysis and other applications.
Delivery information: All our products are shipped with dry ice.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty