Would you like to log in again?
T4 Polynucleotide Kinase (PNK), Intact Genomics
Supplier: Intact Genomics
3232
103753-188EA 213.46 USD InStock
103753-188
T4 Polynucleotide Kinase (PNK), Intact Genomics
Enzymes
T4 polynucleotide kinase catalyzes the transfer and exchange of Pi from the γ position of ATP to the 5'-hydroxyl terminus of polynucleotides (double-and single-stranded DNA and RNA) and nucleoside 3'-monophosphates.
  • Extremely high purity
  • Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
  • 10X T4 PNK reaction buffer
T4 polynucleotide kinase catalyzes the transfer of the γ-phosphate from ATP to the 5'-OH group of single and double-stranded DNAs and RNAs, oligonucleotides or nucleoside 3'-monophosphates. In the presence of ADP, T4 PNK exhibits 5'-phosphatase activity and catalyzes the exchange of phosphate group between 5'-P-oligonucleotides/polynucleotides and ATP. The enzyme is also a 3'-phosphatase.
Delivery information: All our products are shipped on dry ice.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty