Would you like to log in again?
T4 UvsY Protein (from T4 Phage)
Supplier: Intact Genomics
3575 3572
103753-196EA 1292.31 USD InStock
103753-196 103753-194
T4 UvsY Protein (from T4 Phage)
Enzymes
Offers the largest collection of T4 Enzymes and proteins in the industry.
  • Extremely high purity
  • Variety of uses including RPA optimization
  • Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
  • 10X UvsY reaction buffer
UvsY is the phage T4 recombination mediator protein, and structural and biophysical studies provide insights into its role in T4 homologous recombination. During T4 homologous recombination, the UvsX recombinase must compete with the prebound gp32 single-stranded binding protein for DNA-binding sites and T4 UvsY Protein stimulates this filament nucleation event. UvsY is a 15.8 kDa protein with properties that are consistent with its role as mediator; it stimulates the DNA-dependent ATPase activity of UvsX, lowers the critical concentration of UvsX that is required for activity, and promotes strand exchange. UvsY efficiently promotes the UvsX-catalyzed strand invasion reaction by recruiting to ssDNA–gp32 complexes, promotes the release of gp32, and favors the binding of UvsX.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty