Would you like to log in again?
T4 gp32 Protein
Supplier: Intact Genomics
3515
103753-198EA 369.23 USD InStock
103753-198
T4 gp32 Protein
Enzymes
Offers the largest collection of T4 Enzymes and proteins in the industry.
  • Extremely high purity
  • Variety of uses including RPA optimization
  • Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
  • 10X gp32 reaction buffer
T4 gp32 protein is a single-stranded DNA binding protein required for T4 DNA replication, recombination, and repair. It improves the efficiency of reverse transcriptase (RT) during RT-PCR, enhances T4 DNA polymerase activity, as well as increases the yield of PCR product.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty