Would you like to log in again?
Sau DNA Polymerase Large Fragment (from Staphylococcus aureus)
Supplier: Intact Genomics
3595
103753-202EA 575.28 USD InStock
103753-202
Sau DNA Polymerase Large Fragment (from Staphylococcus aureus)
Enzymes
Offers the largest collection of T4 Enzymes and proteins in the industry.
  • Extremely high purity
  • Variety of uses including RPA optimization
  • Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
  • 10X Sau DNA polymerase I reaction buffer
Sau DNA polymerase I, large fragment is a product of the Staphylococcus aureus DNA polymerase I which lacks the N-terminal exonuclease domain (1-293 amino acids). It retains the 5' - 3' polymerase activity of DNA polymerase I but lacks the 5' - 3' exonuclease activity. This large fragment also lacks 3' - 5' exonuclease activity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty