Would you like to log in again?
Tek-Select® Cardboard Cassette Storage Files, IMEB
Supplier: IMEB
Tek-Select®
FC8 FC8-A
103783-040EA 249.23 USD InStock
103783-040 103783-042
Tek-Select® Cardboard Cassette Storage Files, IMEB
Boxes Storage Boxes
Versatile, durable corrugated cardboard boxes are strong yet lightweight, with a moisture-resistant exterior wax coating.
  • Two sets of five drawers
  • Total capacity of 5000 cassettes per case of 10 drawers
  • Strong yet lightweight construction
  • Moisture-resistant with a special wax coating
  • Outer corrugated shell acts as a stabilizer for stacking
  • Available pre-assembled or unassembled
Tek-Select® Cardboard Cassette Storage Files offer a less expensive option to metal and plastic storage filing systems. Boxes can be purchased pre-assembled or unassembled. An outer corrugated shell acts as a stabilizer in keeping units stacked together over a long period of time. Each box section accommodates 500 cassettes. With 10 boxes per case the total capacity of each case is 5000 cassettes.
Accessories information: Optional accessories include four blade types, cooling systems, femoral and small bone holders, splash guards, cleaning stones, and histo-vacuum. The optional cooling system is required for use with diamond saw blades.
Ordering information: Standard blade, mounting board, edge guide, and foot switch included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty