Would you like to log in again?
Tek-Select® Fan-Pad Mini-Formalin Absorbent Pad, IMEB
Supplier: IMEB
FAN-55
103783-056CS 175.38 USD InStock
103783-056
Tek-Select® Fan-Pad Mini-Formalin Absorbent Pad, IMEB
Spill Control Materials
Tek-Select® Fan-Pad mini formalin absorbent are specially treated to absorb formalin and formaldehyde.
  • Pads absorb formalin and formaldehyde
  • Specially treated surfaces
  • 55 sheets per roll
Tek-Select® Fan-Pad mini formalin absorbent pads ensure that formalin products are absorbed and harmful vapors neutralized. Pads can be disposed in regular trash receptacles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty