Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Paraffin, Tek-Select® HP53 for embedding

Supplier: IMEB INC MS
Synonyms: Wax ParaffinParaffin wax

TSHP53
103783-064CS 138.46 USD InStock
103783-064
Paraffin, Tek-Select® HP53 for embedding
Paraffin
Tek-Select® HP53 Embedding Paraffin is a blend of low molecular weight polymers and highly purified paraffin with specialty additives making for reduced tissue compression, resistance and exceptional ribbon continuity.

  • Reduced tissue compression and resistance
  • Exceptional ribbon continuity

HP53 melts uniformly at 52…54 °C. Water bath should be set between 38…42 °C. All Tek-Select paraffin products offer excellent ribboning of tissue sections, even at 2-3 microns. Use TEK-SELECT MM for both processing and embedding with an additive-free formula. Or for high speed production, use TEK-SELECT HP53.

Delivery information: Four 4.4 lb bags per case


Melting Pt: 52…54 °C
CAS Number: 8002-74-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty