Would you like to log in again?
Tek-Select® Bouin’s Fixative Substitutes, IMEB
Supplier: IMEB
Tek-Select®
BFS-32 BFSP3
103783-076EA 218.46 USD InStock
103783-076 103783-078
Tek-Select® Bouin’s Fixative Substitutes, IMEB
Stains Microscopy and staining reagents
A low-hazard replacement for Bouin’s Fixative.
  • Low hazard replacement
  • No picric acid
  • Available in a variety of sizes and pre-filled specimen containers
This substitute provides all the benefits of Bouin’s fixative without the use of picric acid. Available in a variety of sizes and pre-filled specimen containers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty