Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formalin 10% with zinc, Tek-Select®, buffered

Supplier: 0000019204
Synonyms: MethylaldehydeFormolParaformaldehyde aqueous solutionFormaldehyde

BZF5 BZF1011 BZF1 BZF3011
103783-082EA 49.23 USD InStock
103783-082 103783-084 103783-134 103783-124
Formalin 10% with zinc, Tek-Select®, buffered
Formalin
Tek-Select® 10% Buffered Formalin with Zinc is prepared using only high purity chemicals and is ready for application and can be easily used in office-based sites as well as clinics.

  • Ready for application
  • Prepared using only high purity chemicals
  • 10% Formalin solution with 1% Zinc Sulfate and Acetate buffer added in deionized water
  • Acetate buffer stabilizes the pH of solution and prevents the possible formation of formalin pigment

It is recommended as an all-purpose tissue fixative for demonstration of crisp nuclear detail, superior cellular morphology, enhanced Hematoxylin and Eosin staining, special staining and immunohistochemical studies.

It also provides excellent results for lymph nodes and gastric biopsies.

Tek-Select® 10% Buffered Formalin with Zinc presents minimal safety hazards and is non-corrosive.


Formula: CH₂O
MW: 30.03 g/mol
MDL Number: MFCD00003274
CAS Number: 50-00-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty