Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formalin 10% alcoholic solution with zinc, Tek-Select®

Supplier: 0000019204
Danger
Synonyms: MethylaldehydeFormolParaformaldehyde aqueous solutionFormaldehyde

ALZ1011 ALZ5 ALZ3011 ALZ1
103783-128CS 213.85 USD InStock
103783-128 103783-130 103783-132 103783-126
Formalin 10% alcoholic solution with zinc, Tek-Select®
Formalin
Tek-Select® 10% Alcoholic Formalin with Zinc is proprietary compound formulation prepared using high purity chemicals in deionized water and is an excellent alternative for tissue preservation and acts rapidly because it penetrates quickly.

  • Fast dehydration
  • Prepared using high purity chemicals in deionized water
  • Excellent alternative for tissue preservation
  • Acts rapidly because it penetrates quickly

Tek-Select® 10% Alcoholic Formalin with Zinc includes formaldehyde, reagent alcohol, zinc salt, and glacial acetic acid and is an excellent alternative for tissue preservation and acts rapidly because it penetrates quickly.
Dehydration is fast since it can be commenced in absolute alcohol immediately after fixation. Fixation by Alcohols starts at a concentration of 50 – 60% for Ethanol. Tek-Select® 10% Alcoholic Formalin with Zinc contains 85% of Reagent Alcohol, a high concentration for compound fixatives available on the market, without any danger of zinc salt precipitation.

Acetic acid is coagulant for nucleic acids that generally does not fix proteins. It is incorporated in compound fixatives to help prevent the loss of nucleic acids and to counter the shrinkage caused by Alcohol. Acetic acid penetrates tissue very rapidly.

Zinc salts react with a range of tissue end groups including amino, carboxyl and sulphydryl, forming reversible reaction products some of which can be removed with a citrate or EDTA wash. Zinc enhances fixation and staining, particularly of nuclei, in a similar way to mercuric chloride. It has advantages in preserving immuno-reactivity when compared to formalin alone excluding the need for antigen retrieval for some epitopes. Zinc salts are far less toxic than mercury salts and disposal of zinc solutions pose no problem.


Formula: CH₂O
MW: 30.03 g/mol
MDL Number: MFCD00003274
CAS Number: 50-00-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty