Would you like to log in again?
Alpha CrossLaps® ELISA (αCTX-I), Biovendor
Supplier: BioVendor
AC-04F1
103783-142EA 2012.66 USD InStock
103783-142
Alpha CrossLaps® ELISA (αCTX-I), Biovendor
Assays ELISAs
96 wells (1 kit) - ELISA is a sandwich enzyme immunoassay for the quantitative measurement of CTX-I.
The Serum CrossLaps® (CTX-I) ELISA is based on two highly specific monoclonal antibodies against the amino acid sequence of EKAHD-ß-GGR, where the aspartic acid residue (D) is ß-isomerized. In order to obtain a specific signal in the Serum CrossLaps® (CTX-I) ELISA, two chains of EKAHD-ß-GGR must be cross linked. Standards, control, or unknown serum samples are pipetted into the appropriate microtiter wells coated with streptavidin, followed by application of a mixture of a biotinylated antibody and a peroxidase conjugated antibody. Then, a complex between CrossLaps antigens, biotinylated antibody and peroxidase conjugated antibody is generated, and this complex binds to the streptavidin surface via the biotinylated antibody. Following the one step incubation at room temperature, the wells are emptied and washed. A chromogenic substrate is added and the color reaction is stopped with sulfuric acid. Finally, the absorbance is measured.
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty