Would you like to log in again?
Urine BETA CrossLaps® ELISA (β-CTX-I), BioVendor
Supplier: BioVendor
AC-05F1
103783-144EA 1594 USD InStock
103783-144
Urine BETA CrossLaps® ELISA (β-CTX-I), BioVendor
Assays ELISAs
96 wells (1 kit) - ELISA is a sandwich enzyme immunoassay for the quantitative measurement of C-terminal telopeptides of Type l collagen.
The Urine BETA CrossLaps® (CTX-I)ELISA is an enzyme immunological test for the quantification of degradation products of C-terminal telopeptides of type-I collagen in human urine. Summary and explanation of the test type I collagen accounts for more than 90% of the organic matrix of bone and is synthesized primarily in bone (1). During renewal of the skeleton, type I collagen is degraded, and small peptide fragments are excreted into the urine. These fragments can be measured by Urine BETA CrossLaps® (CTX-I)ELISA. Urine BETA CrossLaps® (CTX-I)ELISA demonstrate a high correlation to corresponding measurement of serum samples in Serum CrossLaps® (CTX-I)ELISA. Serum CrossLaps® (CTX-I)ELISA has been reported to be useful for follow-up of anti-resorptive treatment of patients with metabolic bone diseases (3 17).
The Urine BETA CrossLaps® (CTX-I)ELISA is based on two highly specific monoclonal antibodies against the amino acid sequence of EKAHD-β-GGR, where the aspartic acid residue (D) is β-isomerized. In order to obtain a specific signal in the Urine BETA CrossLaps® (CTX-I)ELISA, two chains of EKAHD-β-GGR must be cross-linked. Standards, controls, or unknown urine samples are pipetted into the appropriate microtitre wells coated with streptavidin, followed by application of a mixture of a biotinylated antibody and a peroxidase-conjugated antibody. Then, a complex between CrossLaps antigens, biotinylated antibody and peroxidase-conjugated antibody is generated, and this complex binds to the streptavidin surface via the biotinylated antibody. Following the one-step incubation at room temperature, the wells are emptied and washed. A chromogenic substrate is added and the color reaction is stopped with sulfuric acid. Finally, the absorbance is measured.
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty