Would you like to log in again?
Thiol-Modified Hyaluronan Hydrogel Kit, HyStem®, Advanced BioMatrix
Supplier: Advanced BioMatrix
GS1004F GS311F GS310F
103792-956KT 170 USD InStock
103792-956 103792-924 103792-958
Thiol-Modified Hyaluronan Hydrogel Kit, HyStem®, Advanced BioMatrix
Cell Culture Supplements and Additives
HyStem® Hydrogel Kit - The growth factor delivery matrix
  • Thiolated Hyaluronic Acid Kit
  • "Blank Slate" matrix
  • For first time users, we highly recommend the 2.5 mL or 7.5 mL kit size.
HyStem® Hydrogels - The blank-slate matrix. Completely customizable and xeno-free. Components include Thiol-modified hyaluronan (Gycosil®), Thiol-reactive PEGDA crosslinker (Extralink®), and degassed, deionized water (DG Water).
The HyStem Hydrogel Kits are optimal for culturing stem cells whose natural environment is rich in hyaluronic acid (HA). HyStem hydrogels can be customized by adding extracellular matrix (ECM) proteins or cell attachment peptides into the hydrogel to provide attachment site and/or differentiation signals. They can also be varied by changing the hydrogel rigidity to match that of the native cell environment. The HyStem hydrogel system is viable for cell culture in any format, from T-flasks and tissue culture inserts to 384-well microplates. 
Caution: This product is for R&D use only and is not intended for human or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty