Would you like to log in again?
Thiol-Modified Gelatin, Gelin-S, Advanced BioMatrix
Supplier: Advanced BioMatrix
GS231F-5EA GS230F
103792-940EA 170 USD InStock
103792-940 103792-942
Thiol-Modified Gelatin, Gelin-S, Advanced BioMatrix
Cell Culture Supplements and Additives
Gelin-S is a thiol-modified gelatin that comes as a lyophilized powder. Gelin-S is a component of the HyStem®-C and HyStem®-HP kits, but can be purchased separately here.
  • Thiolated Gelatin
  • Use in conjunction with Extralink and Glycosil for 3D hydrogels
Gelin-S® is thiol-modified gelatin (denatured collagen) and is a component of the HyStem®-C, and HyStem-HP hydrogel kits. Most cells do not grow well on Gelin-S–only hydrogels. Instead, Gelin-S should be used in conjunction with Glycosil® (thiol-modified hyaluronic acid) or Heprasil® (thiol-modified hyaluronic acid with thiol-modified heparin). Reconstituted Gelin-S remains liquid at 15 to 37°C.
The gelatin used to make Gelin-S is from Type A Gelatin, Bloom 250, derived from porcine skin.
Caution: This product is for R&D use only and is not intended for human or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty