Would you like to log in again?
TRIO.BAS™ Satellite Connection Cable
Supplier: Hardy Diagnostics
TRIO.BAS™
BAS272
103792-976EA 360.5 USD InStock
103792-976
TRIO.BAS™ Satellite Connection Cable
Samplers Air Samplers
Satellite cable, 20 meters length (approx. 65') for use with the RABS ISOLATOR and/or MULTIFLEX 1+2 TRIO.BAS instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty