Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

BioPure Gold Nanospheres 1 mg/ml (Au) Cationic/positive surface (BPEI branched polyethylenimine), nanoComposix

Supplier: nanoComposix

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

BIOPURE
AUBB10-30M AUBB100-30M AUBB50-30M
103819-810EA 4027.69 USD InStock
103819-810 103819-812 103819-818
BioPure Gold Nanospheres 1 mg/ml (Au) Cationic/positive surface (BPEI branched polyethylenimine), nanoComposix
Beads Microcarriers
Purified, concentrated, high quality gold nps, sterile and endotoxin-free. BPEI for positive surface.

  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
  • High concentration materials (1 mg/ml, 5.08 mm, 1000 ppm)
  • Extensively purified to remove residual reactants
  • Manufactured sterile; provided in ultra to pure (Milli to Q) DI water
  • BioPure mateirals ideal for in to vitro and in to vivo experiments, nanotoxicology
  • BPEI surface provides a conjugatable amine to terminated positively to charged (cationic) surface

Spherical gold nanoparticles, 1 mg/ml (high concentration), BPEI surface provides a conjugatable amine to terminated positively to charged (cationic) surface. BioPure materials are Sterile and Endotoxin to Free (<5 EU/ml). Precisely engineered, unagglomerated nanospheres with narrow size distributions (CVs <12% for 7 to 15 nm sizes, <15% for 20 to 100 nm sizes, and <18% for 5 nm size).

Each batch extensively characterized to meet highest quality standards, ensuring reliable & reproducible results. All products provided with batch-specific COA's with TEM, DLS, Zeta, UV-Vis measurements.

Colloidal gold has applications in Diagnostics, Lateral Flow Assays, Molecular Detection, Photothermal and Microwave-based Therapies, TEM labels, Nanotoxicology, Plasmonics (SPR / Surface Plasmon Resonance), and novel Color Enhancements and Pigments.

Custom formulations and ISO13485/cGMP compliant materials can be provided on request.

Delivery information: Ships via FedEx Priority Overnight (Domestic). Cold packaging included.
Due to temperature-sensitive nature of these materials, they will only be shipped during the week in order to prevent products from sitting in an uncontrolled temperature environment over the weekend.

Caution: DO NOT FREEZE. Store at 2 to 8 °C and away from light when not in use to maximize shelf life. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty