Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

BioPure Gold Nanospheres 1 mg/ml Citrate (bare/displaceable surface), nanoComposix

Supplier: nanoComposix

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

BIOPURE
AUCB60-30M AUCB100-30M AUCB50-30M AUCB80-30M AUCB20-30M AUCB5-30M AUCB30-30M AUCB40-30M AUCB10-30M
103819-840EA 4027.69 USD InStock
103819-840 103819-832 103819-830 103819-842 103819-836 103819-826 103819-834 103819-824 103819-838
BioPure Gold Nanospheres 1 mg/ml Citrate (bare/displaceable surface), nanoComposix
Beads Microcarriers
Purified, concentrated, high quality gold nps, sterile and endotoxin-free. Bare citrate surface.

  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
  • High concentration materials (1 mg/ml, 5.08 mm, 1000 ppm)
  • Extensively purified to remove residual reactants
  • Manufactured sterile; provided in ultra to pure (Milli to Q) DI water
  • BioPure mateirals ideal for in to vitro and in to vivo experiments, nanotoxicology
  • "Bare" citrate surface is stable and displaceable

Spherical gold nanoparticles, 1 mg/ml (high concentration). Citrate offers stable and readily displaceable surface. BioPure materials are Sterile and Endotoxin to Free (<5 EU/ml). Precisely engineered, unagglomerated nanospheres with narrow size distributions (CVs <12% for 7 to 15 nm sizes, <15% for 20 to 100 nm sizes, and <18% for 5 nm size).

Each batch is extensively characterized to meet highest quality standards, ensuring reliable and reproducible results. All products supplied with COA's including batch-specific TEM, DLS, Zeta, UV-Vis measurements, etc.

Colloidal gold has applications in Diagnostics, Lateral Flow Assays, Molecular Detection, Photothermal and Microwave-based Therapies, TEM labels, Nanotoxicology, Plasmonics (SPR / Surface Plasmon Resonance), and novel Color Enhancements and Pigments.

Custom formulations and ISO13485/cGMP compliant materials can be provided on request.

Delivery information: Ships via FedEx Priority Overnight (Domestic). Cold packaging included.
Due to temperature-sensitive nature of these materials, they will only be shipped during the week in order to prevent products from sitting in an uncontrolled temperature environment over the weekend.

Caution: DO NOT FREEZE. Store at 2 to 8 °C and away from light when not in use to maximize shelf life. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty