Would you like to log in again?
BioReady Gold Nanospheres for Conjugation, nanoComposix
Supplier: nanoComposix
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AUCR40-100M AUCR40-30M AUCR40-5M
103819-866EA 457.69 USD InStock
103819-866 103819-864 103819-868
BioReady Gold Nanospheres for Conjugation, nanoComposix
Beads Microcarriers
40 nm gold nanoparticles for highly reproducible gold conjuates for assays and diagnostics.
  • Achieve reliable gold conjugates for reproducible results in diagnostics and lateral flow assays
  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
BioReady materials are engineered for reliable conjugation to antibodies, proteins, peptides, and other biomolecules. Ideal for lateral flow and other detection and diagnostics.
Each batch is extensively characterized to meet highest quality standards, ensuring reliable & reproducible results. All products supplied with batch-specific COA's with TEM, DLS, Zeta, & UV-Vis.
Colloidal gold has applications in Diagnostics, Lateral Flow Assays, Molecular Detection, Photothermal and Microwave-based Therapies, TEM labels, Nanotoxicology, Plasmonics (SPR / Surface Plasmon Resonance), and novel Color Enhancements and Pigments.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.
Delivery information: Ships at ambient temperatures
Caution: DO NOT FREEZE. Store at 2 to 8 °C when not in use to maximize shelf life. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty