Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

BioPure Gold Nanospheres 1 mg/ml (Au) PVP (polyvinylpyrrolidone for high stability), nanoComposix

Supplier: nanoComposix

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

BIOPURE
AUPB50-30M AUPB10-30M AUPB100-30M
103819-950EA 4027.69 USD InStock
103819-950 103819-942 103819-944
BioPure Gold Nanospheres 1 mg/ml (Au) PVP (polyvinylpyrrolidone for high stability), nanoComposix
Beads Microcarriers
Purified, concentrated, high quality gold nps, sterile and endotoxin-free. PVP for stability.

  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
  • High concentration materials (1 mg/ml, 5.08 mm, 1000 ppm)
  • Extensively purified to remove residual reactants
  • Manufactured sterile; provided in ultra to pure (Milli to Q) DI water
  • BioPure mateirals ideal for in to vitro and in to vivo experiments, nanotoxicology
  • PVP surface provides strong steric stability

Spherical gold nanoparticles, 1 mg/ml (high concentration), PVP polymer surface provides strong steric stability (40 kD polyvinylpyrrolidone). BioPure materials are Sterile and Endotoxin to Free (<5 EU/ml). Precisely engineered, unagglomerated nanospheres with narrow size distributions (CVs <12% for 7 to 15 nm sizes, <15% for 20 to 100 nm sizes, and <18% for 5 nm size).

Each batch is extensively characterized to meet highest quality standards, ensuring reliable & reproducible results. All products supplied with COA's including batch-specific TEM, DLS, Zeta, UV-Vis measurements, etc.

Colloidal gold has applications in Diagnostics, Lateral Flow Assays, Molecular Detection, Photothermal and Microwave-based Therapies, TEM labels, Nanotoxicology, Plasmonics (SPR / Surface Plasmon Resonance), and novel Color Enhancements and Pigments.

Custom formulations and ISO13485/cGMP compliant materials can be provided on request

Delivery information: Ships at ambient temperatures

Caution: DO NOT FREEZE. Store at 2 to 8 °C when not in use to maximize shelf life. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty